Learn More
PDC Meditech Direct Thermal IR Paper Label, 1000 Labels/Roll, 2 Rolls/Case
Catalog No. 21235794
Change view
Click to view available options
Quantity:
1000 Labels/Roll, 2 Rolls/Cs.
Catalog No. Quantity
21-235-794 1000 Labels/Roll, 2 Rolls/Cs.
1 options
Catalog No. 21-235-794 Supplier PDC Supplier No. LDWHP123
  • $215.00
    $160.65 / Case of 2
    Save $54.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
Durable labels with permanent adhesive, compatible with IR printing, designed for reliable specimen and patient identification in healthcare.
  • Designed for use with Meditech systems to ensure accurate clinical workflows
  • Permanent adhesive keeps labels securely attached to vials, tubes, and containers
  • White face stock provides clear, legible printing for patient and specimen information
  • Supports infrared (IR) printing for system compatibility and reliable data capture
  • Core design fits standard dispensers for easy application
  • Bulk packaging provides sufficient supply for high-volume healthcare labeling needs
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1000 Labels/Roll, 2 Rolls/Cs.
Dimensions (L x W) 3 3/8 x 5 1/4 in.
Length (English) 3.375 in.
Width (English) 5.25 in.
Width (Metric) 133.35 mm