Learn More
PDC Meditech Direct Thermal IR Paper Label, White
Catalog No. 21235809
Change view
Click to view available options
Quantity:
3600 Labels/Roll
Catalog No. Quantity
21-235-809 3600 Labels/Roll
1 options
Catalog No. 21-235-809 Supplier PDC Supplier No. LMED51
Add to Cart
Add to Cart
Permanent adhesive labels with infrared-sensitive material, designed for reliable specimen and patient identification in clinical settings.
  • PDC infrared-sensitive labels designed for accurate clinical and laboratory workflows
  • Permanent adhesive ensures secure attachment to vials, tubes, and containers
  • White face stock provides clear, legible printing for patient and specimen information
  • Compatible with standard dispensers for convenient application
  • High-volume labeling solution supports efficient workflow and inventory management
  • Durable material improves print longevity and scanning reliability in healthcare environments
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 3600 Labels/Roll
Dimensions (L x W) 4 x 1 1/4 in.
Length (English) 4 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm