Learn More
PDC Meditech Direct Thermal Paper Label, 1000 Labels/Roll, 8 Rolls/Case
Catalog No. 21235803
Change view
Click to view available options
Includes:
Wristband - Wound In
Wristband - Wound Out
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
21-235-803 Wristband - Wound In
21-235-798 Wristband - Wound Out
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-235-803 Supplier PDC Supplier No. LDWHP58
Add to Cart
Add to Cart
Strong adhesive labels designed for accurate identification in healthcare and clinical labeling tasks.
  • Permanent adhesive provides secure attachment for reliable use in clinical settings
  • White stock delivers clear, easy-to-read printing for patient and specimen information
  • Label size 4 in. x 1 1/4 in. gives enough space for detailed identification
  • 1 in. core works well with compact dispensers and printers for smooth handling
  • Ideal choice for hospitals, laboratories, and other healthcare facilities
  • Each roll has 1000 labels and cases include 8 rolls to support high-volume needs
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Includes Wristband - Wound In
Quantity 1000 Labels/Roll, 8 Rolls/Cs.
Dimensions (L x W) 4 x 1 1/4 in.
Length (English) 4 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm