Learn More
PDC Meditech Direct Thermal Paper Label, 500 Labels/Roll
Catalog No. 21235799
Change view
Click to view available options
Quantity:
500 Labels/Roll
Catalog No. Quantity
21-235-799 500 Labels/Roll
1 options
Catalog No. 21-235-799 Supplier PDC Supplier No. LDWHP165
Add to Cart
Add to Cart
Reliable thermal labels designed for healthcare, ensuring durable adhesion and clear identification for patient and specimen use.
  • Permanent adhesive for secure labeling
  • Sharp, legible print on white face stock
  • Suitable for specimen and patient information
  • Compatible with compact printers and dispensers
  • Ensures durable and reliable labeling in healthcare
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 500 Labels/Roll
Dimensions (L x W) 3 3/8 x 5 1/4 in.
Length (English) 3.375 in.
Width (English) 5.25 in.
Width (Metric) 133.35 mm