Learn More
PDC Meditech Direct Thermal Paper Label, White with Red, 1000 Labels/Roll, 8 Rolls/Case
Catalog No. 21235804
Change view
Click to view available options
Quantity:
1000 Labels/Roll, 8 Rolls/Cs.
Catalog No. Quantity
21-235-804 1000 Labels/Roll, 8 Rolls/Cs.
1 options
Catalog No. 21-235-804 Supplier PDC Supplier No. LDWHP63
Add to Cart
Add to Cart
Strong adhesive labels in white with red markings, designed for reliable use in healthcare and clinical labeling tasks.
  • Permanent adhesive ensures labels stay firmly attached in medical and lab use
  • White face stock with red markings enhances visibility and identification
  • Label size 4 in. x 1 1/4 in. provides enough space for specimen and patient details
  • 1 in. core fits standard dispensers for easy application
  • Packaged with 1000 labels per roll and 8 rolls per case for bulk supply
  • Widely used in hospitals, laboratories, and other healthcare facilities
Specifications
Adhesive Type Permanent
Coating None
Color White, Red Border
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 1000 Labels/Roll, 8 Rolls/Cs.
Dimensions (L x W) 4 x 1 1/4 in.
Length (English) 4 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm