Learn More
PDC Meditech Direct Thermal Paper Label, White with Red, 3600 Labels/Roll
Catalog No. 21235806
Change view
Click to view available options
Quantity:
3600 Labels/Roll
Catalog No. Quantity
21-235-806 3600 Labels/Roll
1 options
Catalog No. 21-235-806 Supplier PDC Supplier No. LDWHP67
Add to Cart
Add to Cart
Strong adhesive labels with red markings on white stock, designed for clear identification in healthcare and clinical labeling.
  • Permanently adhesive surface ensures labels remain secure on clinical and lab items
  • White background with red markings improves visibility for fast recognition
  • Label size 4 in. x 1 1/4 in. offers enough space for specimen and patient details
  • 3 in. core fits larger dispensers for high-volume labeling needs
  • Supplied with 3600 labels per roll to support extended and frequent use
  • Trusted for hospitals, laboratories, and other healthcare environments
Specifications
Adhesive Type Permanent
Coating None
Color White, Red Border
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 3600 Labels/Roll
Dimensions (L x W) 4 x 1 1/4 in.
Length (English) 4 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm