Learn More
PDC Meditech Direct Thermal Paper Permanent, 3600 Labels/Roll
Catalog No. 21235838
Change view
Click to view available options
Quantity:
3600 Labels/Roll
Catalog No. Quantity
21-235-838 3600 Labels/Roll
1 options
Catalog No. 21-235-838 Supplier PDC Supplier No. TDMIT3
  • $89.00
    $66.65 / Case of 3600
    Save $22.35
    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart
High-quality direct thermal labels compatible with Meditech, featuring permanent adhesive for clear, durable specimen labeling
  • Direct thermal label for Meditech systems
  • Size ensures clear and legible prints
  • Permanent adhesive for reliable label retention
  • Core fits standard thermal printers
  • White paper offers high contrast for readability
  • High-volume capacity for efficient labeling
  • Easy inventory management
  • No ink or toner needed for cost savings
  • Smudge-resistant surface protects printed data
  • Ideal for healthcare specimen and sample labeling
Specifications
Adhesive Type Permanent
Coating None
Color White
Finish Matte
Format Label
Material Paper
Printable Sides Front
Print Technology Direct Thermal
Product Type Label
Sterility Non-sterile
Quantity 3600 Labels/Roll
Dimensions (L x W) 4 x 1 1/4 in.
Length (English) 4 in.
Width (English) 1.25 in.
Width (Metric) 31.75 mm