Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Medium 154CF/PRF, Kit
GREENER_CHOICE

Catalog No. M154CFPRF50
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M154CFPRF50 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M154CFPRF50 Supplier Gibco™ Supplier No. M154CFPRF500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Medium 154CF/PRF, Kit

A sterile, liquid medium for the culture of human epidermal keratinocytes. Medium 154CF/PRF is Medium 154 prepared without calcium chloride or phenol red. Calcium chloride is provided as a separate component with each bottle of medium. This basal medium requires the addition of HKGS (S-001-5) or HKGS Kit (S-001-K) prior to use.

TRUSTED_SUSTAINABILITY

Specifications

Cell Type Keratinocyte Cells
Classification Serum-free
Form Liquid
Product Type Medium 154CF Kit
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
Without Additives No Phenol Red
Product Line Cascade Biologics
Quantity 500 mL
Shipping Condition Room Temperature
Content And Storage Store in refrigerator (2–8°C) and protect from light.
Culture Type Mammalian Cell Culture
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.