Learn More
Sigma Aldrich Fine Chemicals Biosciences Medium 199, 1 L
Catalog No. NC0862677
Encompass
Originally developed as a completely defined media formulation for the culture of primary explants. These media have broad applicability, particularly for non-transformed cells and are widely used for vaccine production, primary pancreatic explants, and lens tissues.
Catalog No. NC0862677 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M45301L
Originally developed as a completely defined media formulation for the culture of primary explants. These media have broad applicability, particularly for non-transformed cells and are widely used for vaccine production, primary pancreatic explants, and lens tissues.