Learn More
Sigma Aldrich Fine Chemicals Biosciences Medium 199 With Earle's salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | 500ML
Catalog No. NC2061727
Encompass
Medium 199 With Earle's salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | 500ML
Catalog No. NC2061727 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M4530500ML
Medium 199 With Earle's salts, L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture | 500ML