Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Medium 199 with Earle's Salt (E199), with L-glutamine, without Sodium Bicarbonate

Catalog No. 091020122 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
09-102-0122 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-102-0122 Supplier MP Biomedicals™ Supplier No. 091020122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Medium 199 with Earle's Salt (E199), with L-glutamine, without Sodium Bicarbonate is a completely defined nutritional source for cell culture.

  • Medium 199, originally described by Morgan and his colleagues (1950), is a completely defined nutritional source for cell culture.
  • Their investigations demonstrated that cell growth could be measured in this medium.
  • It has broad species applicability including the culturing of non-transformed cell types.
  • It may be used for vaccine production and the in vitro cultivation of rat lens tissues and primary mouse pancreatic epithelial explants.
  • It is available in either Earle's or Hanks' salts.
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Quantity 10 L

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.