Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Medium 199 (modified) with Hanks' Salts with Glutamine without Sodium Bicarbonate

Catalog No. MP091023122 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 x 10 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP091023122 1 x 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MP091023122 Supplier MP Biomedicals™ Supplier No. 091023122
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Medium 199 (modified) with Hanks' Salts with Glutamine without Sodium Bicarbonate

  • H199 Powder
  • Hanks' salts with L-glutamine
  • Without sodium bicarbonate
TRUSTED_SUSTAINABILITY

Specifications

Form Powder
Product Type Cell Culture Media
Quantity 1 x 10 L
Content And Storage Store at 0-5C
Description Hanks' salts with L-glutamineWithout sodium bicarbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.