Learn More
Doccol Corporation Medium A MCAO suture 60L1012C
SDP
Supplier:  Doccol Corporation 60231012PK10
Encompass
Key Features:1. Filament Specifications: The 6-0 filament size has a diameter ranging from 0.09 to 0.11 mm and a length of 20 mm, providing superb pushability for easy handling and excellent flexibility for reducing impact on Willis Circle.2. Silicone Rubber Coating: With a diameter of 0.23 +/- 0.02 mm, the silicone rubber coating ensures smooth and controlled insertion, enhancing precision in your experiments. Such coating provides ideal dimensions for MCAO procedures in mice within the body weight range of 30 +/- 5 grams. The coating extends 10-12 mm, catering to the needs of permanent MCAO or transient (less than 60 min) MCAO followed by reperfusion.3. Suitable for Highly Skilled Surgeons and for MCAO models that requires the highest consistence of infarct volume: The 10-12 mm coating length is optimized for providing the highest consistence and reliability of infarct volume.
Catalog No. NC3243367