Learn More
Memmert Memmert Humidity Chamber HCP 105, 230V +18C to +90C, 20-95%RH- 40mm silicone entry port with stopper rear wall
SDP
Catalog No. 5000451139
Encompass_Preferred
Memmert Humidity Chamber HCP 105, 230V +18C to +90C, 20-95%RH- 40mm silicone entry port with stopper rear wall
Catalog No. 50-004-51139 Supplier Memmert Supplier No. HCP 105 230V
Memmert Humidity Chamber HCP 105, 230V +18C to +90C, 20-95%RH- 40mm silicone entry port with stopper rear wall