Learn More
Chemglass Life Sciences Clamp, Three Prong Extension, Medium, Dual Adjust, 1/2" OD x 12" Arm
SDP
Catalog No. CG9205E10
Encompass_Preferred
Dual adjust nickel-plated zinc alloy clamp has high tensile strength and corrosion resistance. Fully enclosed stainless steel springs provide smooth operation. Extension arm for both sizes is 1/2" (13mm) OD by 12" Long. Complete wtih removable vinyl and fiberglass covers for easy cleaning.
Catalog No. CG9205E10 Supplier Chemglass Life Sciences Supplier No. CG9205E10
Dual adjust nickel-plated zinc alloy clamp has high tensile strength and corrosion resistance. Fully enclosed stainless steel springs provide smooth operation. Extension arm for both sizes is 1/2" (13mm) OD by 12" Long. Complete wtih removable vinyl and fiberglass covers for easy cleaning.