Learn More
Memmert Memmert CO2 Incubator ICO 150, 230Volt, +5C above Ambient to +50C, Humidity, CO2 0 to 20%,
SDP
Catalog No. 5000451069
Encompass_Preferred
Memmert CO2 Incubator ICO 150, 230Volt, +5C above Ambient to +50C, Humidity, CO2 0 to 20%,
Catalog No. 50-004-51069 Supplier Memmert Supplier No. ICO 150 230V
Memmert CO2 Incubator ICO 150, 230Volt, +5C above Ambient to +50C, Humidity, CO2 0 to 20%,