Learn More
Memmert Memmert CO2 Incubator ICO 105, 230Volt, +5C above Ambient to +50C, Humidity, CO2 0 to 20%,
SDP
Catalog No. 5000451102
Encompass_Preferred
Memmert CO2 Incubator ICO 105, 230Volt, +5C above Ambient to +50C, Humidity, CO2 0 to 20%,
Catalog No. 50-004-51102 Supplier Memmert Supplier No. ICO 105 230V
Memmert CO2 Incubator ICO 105, 230Volt, +5C above Ambient to +50C, Humidity, CO2 0 to 20%,