Learn More
Chemgenes Corporation Medium Drying Trap, 10 PK, suitable for oligonucleotide synthesis, 50-100 mL acetonitrile solution, Reduces moisture <50 ppm, Fully activated
SDP
Catalog No. NC1738849
Encompass
Medium Drying Trap, 10 PK, suitable for oligonucleotide synthesis, 50-100 mL acetonitrile solution, Reduces moisture <50 ppm, Fully activated
This item is not returnable. View return policy
Catalog No. NC1738849 Supplier Chemgenes Corporation Supplier No. DMT1974
Medium Drying Trap, 10 PK, suitable for oligonucleotide synthesis, 50-100 mL acetonitrile solution, Reduces moisture <50 ppm, Fully activated