Learn More
MSC The Fairbanks Company Swivel Top Plate Caster, Rubber, 170 Lb Capacity, 3 in. Diameter, 1 ea.
Catalog No. 5000516978
Encompass
This medium-duty swivel top plate caster is designed for reliable mobility and durability. It features a 3 in. diameter rubber wheel with a roller bearing to facilitate smooth operation and efficient load handling in various applications.
  • Features double ball race swivels for improved performance
  • Includes a large secondary raceway for enhanced swiveling, efficiency, and shock absorption
  • Constructed with a 3-1/8 in. x 4-1/8 in. top plate mount
  • Offers a 170 lb load capacity
  • Rubber wheel construction with a 1-3/8 in. wheel width
  • Finished with zinc coating
Catalog No. 50-005-16978 Supplier MSC Supplier No. 01357086
This medium-duty swivel top plate caster is designed for reliable mobility and durability. It features a 3 in. diameter rubber wheel with a roller bearing to facilitate smooth operation and efficient load handling in various applications.
  • Features double ball race swivels for improved performance
  • Includes a large secondary raceway for enhanced swiveling, efficiency, and shock absorption
  • Constructed with a 3-1/8 in. x 4-1/8 in. top plate mount
  • Offers a 170 lb load capacity
  • Rubber wheel construction with a 1-3/8 in. wheel width
  • Finished with zinc coating