Learn More
Electron Microscopy Sciences SEM Medium Resolution Gold on Carbon Test Specimen, ISI Stub
SDP
Catalog No. 501931211
Encompass_Preferred
Larger gold particles in this test with wider gaps, allow for structured microscope setup. Magnification range is 800-10,000x.
Type: ISI Stub
Catalog No. 50-193-1211 Supplier Electron Microscopy Sciences Supplier No. 7951603
Larger gold particles in this test with wider gaps, allow for structured microscope setup. Magnification range is 800-10,000x.
Type: ISI Stub