Learn More
Electron Microscopy Sciences SEM Medium Resolution Au on Carbon Test Specimen, 12.5mm Pin Stub
SDP
Catalog No. 501931209
Encompass_Preferred
Larger gold particles in this test with wider gaps, allow for structured microscope setup. Magnification range is 800-10,000x.
Type: 12.5mm Pin Stub
Catalog No. 50-193-1209 Supplier Electron Microscopy Sciences Supplier No. 7951601
Larger gold particles in this test with wider gaps, allow for structured microscope setup. Magnification range is 800-10,000x.
Type: 12.5mm Pin Stub