Learn More
Fine Science Tools Strabismus Scissors, Stainless Steel, 11.5 cm
SDP
Catalog No. NC9410793
Encompass
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 23mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No
Catalog No. NC9410793 Supplier Fine Science Tools Supplier No. 1407411
Add to Cart
Add to Cart
Tip Shape: Straight/ Tips: Blunt-Blunt/ Cutting Edge: 23mm/ Alloy / Material: Stainless Steel/ Length: 11.5cm/ Serrated: No