Learn More
Memmert Memmert Climate Chamber ICH 260L, 115V, light module acc ICH Q1B option 2, light intensity 8,000Lux, 4 Fluorescent lights with cold white light - 2 fluorescent lights with UV lamps. Temp range -10C to +60C without humidity, with humidity 0-60C, RH 10-80%
SDP
Catalog No. 5000451104
Encompass_Preferred
Memmert Climate Chamber ICH 260L, 115V, light module acc ICH Q1B option 2, light intensity 8,000Lux, 4 Fluorescent lights with cold white light - 2 fluorescent lights with UV lamps. Temp range -10C to +60C without humidity, with humidity 0-60C, RH 10-80%
Catalog No. 50-004-51104 Supplier Memmert Supplier No. ICH 260L 115V
Memmert Climate Chamber ICH 260L, 115V, light module acc ICH Q1B option 2, light intensity 8,000Lux, 4 Fluorescent lights with cold white light - 2 fluorescent lights with UV lamps. Temp range -10C to +60C without humidity, with humidity 0-60C, RH 10-80%