Learn More
Memmert Memmert Climate ChamberICH 110 L, 115v 50/60Hz. 0-60C, 10-60 with humidity and/or light, 10-80%RH, Illumination acc. ICH Q1B, option 2; separately switchable via controller, one box; 3 fluorescent lights with cold white light. Light color 865 6,500 K; 2 fluo
SDP
Catalog No. 5000451057
Encompass_Preferred
Memmert Climate ChamberICH 110 L, 115v 50/60Hz. 0-60C, 10-60 with humidity and/or light, 10-80%RH, Illumination acc. ICH Q1B, option 2; separately switchable via controller, one box; 3 fluorescent lights with cold white light. Light color 865 6,500 K; 2 fluo
Catalog No. 50-004-51057 Supplier Memmert Supplier No. ICH 110L 115V
Memmert Climate ChamberICH 110 L, 115v 50/60Hz. 0-60C, 10-60 with humidity and/or light, 10-80%RH, Illumination acc. ICH Q1B, option 2; separately switchable via controller, one box; 3 fluorescent lights with cold white light. Light color 865 6,500 K; 2 fluo