Learn More
Ampco Medium Valve Wheel Hook
SDP
Catalog No. 19175145 Shop All Ampco Metal Inc. Products
Change view
Click to view available options
Length (English):
19 -3/4 in.
Catalog No. Length (English)
19-175-145 19 -3/4 in.
1 options
Catalog No. 19-175-145 Supplier Ampco Supplier No. WV38
Add to Cart
Add to Cart
Non-sparking, non-magnetic, and corrosion-resistant valve wheel hook. Made in the USA.
  • F-type valve wheel hook
  • Aluminum bronze material, beryllium free.
  • Includes lifetime warranty.
  • Please see image for more information regarding measurements.
  • 19 and 3/4 in. length.
  • a (jaw opening size): 2 in.
  • b (throat depth): 3 and 7/16 in.
Specifications
Product Type Valve Wheel Hook
Length (English) 19 -3/4 in.
Quantity 1 Ea.