Learn More
Andwin Scientific MEDIUM WEIGH BOATS CS/500
SDP
Catalog No. NC9148527
Encompass_Preferred
The Antistatic Polystyrene Weigh Boats are idea toolsl for weighing liquid or powdered samples in the laboratory. This product has a a smooth surface that gives accurate pour-outs with minimal sample loss and facilitates weighing of static-affected samples. The weigh boats are versatile, durable, economical, and secure. This product is sold in a case of 500.
Catalog No. NC9148527 Supplier Andwin Scientific Supplier No. 12577027
The Antistatic Polystyrene Weigh Boats are idea toolsl for weighing liquid or powdered samples in the laboratory. This product has a a smooth surface that gives accurate pour-outs with minimal sample loss and facilitates weighing of static-affected samples. The weigh boats are versatile, durable, economical, and secure. This product is sold in a case of 500.