Learn More
Medix™ Ery-Tic RBC Test Kit
Catalog No. 23230232
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
23-230-232 1 Kit
1 options
This item is not returnable. View return policy
Catalog No. 23-230-232 Supplier Medix™ Supplier No. BEP4012
Add to Cart
Add to Cart
This item is not returnable. View return policy
For microscopic counting of erythrocytes (red blood cells)
  • Replacement for Unopette
  • Includes capillaries
Specifications
Type Ery-Tic RBC Test Kit
Disposable Single-use
Includes Capillaries, premeasured, prefilled reagent reservoirs
Intended Use Microscopic counting of Erythrocytes (RBC)
Quantity 1 Kit