Learn More
Medical Chemical Corporation Medizime Prime, Tri-enzyme detergent cleaning, low-foaming, concentrated formula for manual or automated systems
SDP
Catalog No. NC3934481
Encompass_Preferred
Medizime Prime is a concentrated, low-foaming tri-enzyme detergent formulated for the effective removal of proteinaceous soils, blood, lipids, and organic debris from medical and laboratory instruments. Engineered for use in manual cleaning, ultrasonic units, evacuation systems, and automated washers, it delivers rapid soil breakdown while maintaining broad material compatibility.
Its triple-enzyme blend supports efficient degradation of complex organic contaminants, helping improve cleaning performance prior to high-level disinfection or sterilization processes. The neutral pH (6.4) formulation is safe for use on metal, glass, rubber, plastics, and flexible instrumentation.
Highly concentrated at a 1 oz per gallon dilution, Medizime Prime supports cost-effective reprocessing with reduced residue and low foam generation.
Catalog No. NC3934481 Supplier Medical Chemical Corporation Supplier No. 3350PR1GL.
Medizime Prime is a concentrated, low-foaming tri-enzyme detergent formulated for the effective removal of proteinaceous soils, blood, lipids, and organic debris from medical and laboratory instruments. Engineered for use in manual cleaning, ultrasonic units, evacuation systems, and automated washers, it delivers rapid soil breakdown while maintaining broad material compatibility.
Its triple-enzyme blend supports efficient degradation of complex organic contaminants, helping improve cleaning performance prior to high-level disinfection or sterilization processes. The neutral pH (6.4) formulation is safe for use on metal, glass, rubber, plastics, and flexible instrumentation.
Highly concentrated at a 1 oz per gallon dilution, Medizime Prime supports cost-effective reprocessing with reduced residue and low foam generation.