Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medline™ Avant Gauze

Catalog No. 23666040 Shop All Medline Industries Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
8000 Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-666-040 8000 Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 23-666-040 Supplier Medline™ Supplier No. NON25224
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Material Rayon, Polyester
Product Type Gauze Sponge
Sterility Non-sterile
Length (English) 2 in.
Length (Metric) 5.08 cm
Width (Metric) 5.08 cm
Width (English) 2 in.
Quantity 8000 Cs.
Dimensions (L x W) 2 x 2 in.
For Use With (Application) Produces less lint and is less traumatic to fragile granulating wound tissue, significantly faster wicking ability which promotes a healthier healing environment
Thickness 4-Ply
Absorbancy Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.