Learn More
Enterprise Technology Solutions Medline Bandages,Self,Adhesive,Plastic,3/4"x3",100/BX,BN
SDP
Catalog No. 5000827841
Encompass_Preferred
Made with durable polyethylene and perforated for breathability, Curad Plastic Adhesive Bandages provide comfortable, everyday, long-lasting wound protection. Four-sided seal helps keep dirt and germs out for an optimal healing environment. Plastic features air holes for breathability while absorbent, nonstick island pad provides absorbency. Rounded edges allow great fit and contour. Sterile bandages are not made with natural rubber latex.
Catalog No. 50-008-27841 Supplier Enterprise Technology Solutions Supplier No. MIINON25500
Made with durable polyethylene and perforated for breathability, Curad Plastic Adhesive Bandages provide comfortable, everyday, long-lasting wound protection. Four-sided seal helps keep dirt and germs out for an optimal healing environment. Plastic features air holes for breathability while absorbent, nonstick island pad provides absorbency. Rounded edges allow great fit and contour. Sterile bandages are not made with natural rubber latex.