Learn More
Enterprise Technology Solutions Medline Earplugs,High-Fidelity Foam,50PR/BG,OE
SDP
Supplier:  Enterprise Technology Solutions MIIMDSEARPLUG
Encompass_Preferred
Earplugs are made of high-fidelity foam to reduce noise up to 35 decibels. Highly visible orange earplugs protect your ears from noise by reducing the noise level without completely blocking out the sound. Each pair comes individually wrapped.
Catalog No. 50-008-28187