Learn More
Enterprise Technology Solutions Medline Exam Gloves, Powder-Free, X-Large, 230/BX, BE
SDP
Catalog No. 5000828369
Encompass_Preferred
Powder-free exam gloves are made using the latest advancements in technology to offer outstanding comfort and performance. Durable nitrile material is incredibly soft while providing excellent chemical-resistance and barrier protection. Latex-free gloves are tested for safe use with chemotherapy drugs. Enhanced tactile sensitivity allows you to accurately work with smaller items. Extra-large gloves are recommended for use in areas that may involve higher levels of stress on exam gloves or areas that deal with harsh chemicals or liquid immersion.
Catalog No. 50-008-28369 Supplier Enterprise Technology Solutions Supplier No. MIIMDS6804
Powder-free exam gloves are made using the latest advancements in technology to offer outstanding comfort and performance. Durable nitrile material is incredibly soft while providing excellent chemical-resistance and barrier protection. Latex-free gloves are tested for safe use with chemotherapy drugs. Enhanced tactile sensitivity allows you to accurately work with smaller items. Extra-large gloves are recommended for use in areas that may involve higher levels of stress on exam gloves or areas that deal with harsh chemicals or liquid immersion.