Learn More
Enterprise Technology Solutions Medline Exam Gloves, Vinyl, Powder Free, Large, 150/BX, Clear
SDP
Catalog No. 5000828364
Encompass_Preferred
Powder-free, vinyl hand gloves are a cost-effective alternative that's perfect for general patient care. Latex-free construction uses advanced technology to dramatically improve tactile sensitivity and durability for efficient use. Ambidextrous design allows each glove to be used for either hand. Gloves are designed for use in noncritical care units, laboratories and more. This product has an ingredient identified by California Proposition 65.
Catalog No. 50-008-28364 Supplier Enterprise Technology Solutions Supplier No. MII6MSV513
Powder-free, vinyl hand gloves are a cost-effective alternative that's perfect for general patient care. Latex-free construction uses advanced technology to dramatically improve tactile sensitivity and durability for efficient use. Ambidextrous design allows each glove to be used for either hand. Gloves are designed for use in noncritical care units, laboratories and more. This product has an ingredient identified by California Proposition 65.