Learn More
Enterprise Technology Solutions Medline Eyewash,Refill,Saline,32 oz,CL
SDP
Catalog No. 5000828457
Encompass_Preferred
Eyewash refill contains Eyesaline, a preserved and buffered saline solution physiologically balanced to the pH and salinity of human tears. While not intended to replace primary eyewash equipment, it is an important supplement for immediate treatment before the injured person is able to reach primary eyewash equipment. It features an extended-flow nozzle and integral gripping surface for convenience.
Catalog No. 50-008-28457 Supplier Enterprise Technology Solutions Supplier No. MIISPV3204550
Eyewash refill contains Eyesaline, a preserved and buffered saline solution physiologically balanced to the pH and salinity of human tears. While not intended to replace primary eyewash equipment, it is an important supplement for immediate treatment before the injured person is able to reach primary eyewash equipment. It features an extended-flow nozzle and integral gripping surface for convenience.