Learn More
Enterprise Technology Solutions Medline Eyewash Station,Wall-Mount,DBL,32ozBttle,25"x9"x15",Green
SDP
Catalog No. 5000827903
Encompass_Preferred
Eyesaline eyewash stations deliver emergency flushing and decontamination until primary emergency eyewash devices become available. Durable, open-face design holds bottles firmly in place while still providing easy access. Tamper-resistant bottles feature extended-flow openings that provide longer flow rates than conventional bottles. Easily mount wall stations in optimal locations for convenient access (mounting fasteners included). Integral overhang protects bottles from dust and debris to minimize maintenance requirements.
Catalog No. 50-008-27903 Supplier Enterprise Technology Solutions Supplier No. MIISPV4620000
Eyesaline eyewash stations deliver emergency flushing and decontamination until primary emergency eyewash devices become available. Durable, open-face design holds bottles firmly in place while still providing easy access. Tamper-resistant bottles feature extended-flow openings that provide longer flow rates than conventional bottles. Easily mount wall stations in optimal locations for convenient access (mounting fasteners included). Integral overhang protects bottles from dust and debris to minimize maintenance requirements.