Learn More
Enterprise Technology Solutions Medline Face Masks, w/Earloops, Individual Wrapped, 10/BX, BE/WE
SDP
Catalog No. 5000828201
Encompass_Preferred
Medical-grade FaceMasks offer maximum barrier protection with 99 percent bacterial filtration. Slip on easily with built-in earloops. Noseguard is adjustable for a comfortable fit. Anti-fog foam strip helps with fogging glasses. Single-use face masks are all fluid-resistant. They are not made with natural rubber latex for safer use by those with latex allergies. Each one is individually wrapped for sanitary use.
Catalog No. 50-008-28201 Supplier Enterprise Technology Solutions Supplier No. MIICUR812S
Medical-grade FaceMasks offer maximum barrier protection with 99 percent bacterial filtration. Slip on easily with built-in earloops. Noseguard is adjustable for a comfortable fit. Anti-fog foam strip helps with fogging glasses. Single-use face masks are all fluid-resistant. They are not made with natural rubber latex for safer use by those with latex allergies. Each one is individually wrapped for sanitary use.