Learn More
Enterprise Technology Solutions Medline Face Shields,FullLength,Disposable,Antifog,24/BX,CL
SDP
Catalog No. 5000827949
Encompass_Preferred
Disposable Full-Length Face Shields provide anti-fog protection from splashing and spraying. Lightweight design features a foam band across your forehead and an elastic band that keeps the shield secure. Optically clear polyester allows maximum visibility. Wear them with or without glasses.
Catalog No. 50-008-27949 Supplier Enterprise Technology Solutions Supplier No. MIINONFS300
Disposable Full-Length Face Shields provide anti-fog protection from splashing and spraying. Lightweight design features a foam band across your forehead and an elastic band that keeps the shield secure. Optically clear polyester allows maximum visibility. Wear them with or without glasses.