Learn More
Enterprise Technology Solutions Medline Face Shields,FullLength,ExtraLong,Dispsable,Antifog,40/CT,CL
SDP
Catalog No. 5000827939
Encompass_Preferred
Disposable Full-Length Extra-Long Face Shields provide anti-fog protection from splashing and spraying. Lightweight design features a foam band across your forehead and an elastic band that keep the shield secure. Optically clear polyester allows maximum visibility. Wear them with or without glasses.
Catalog No. 50-008-27939 Supplier Enterprise Technology Solutions Supplier No. MIINONFS100
Disposable Full-Length Extra-Long Face Shields provide anti-fog protection from splashing and spraying. Lightweight design features a foam band across your forehead and an elastic band that keep the shield secure. Optically clear polyester allows maximum visibility. Wear them with or without glasses.