Learn More
Enterprise Technology Solutions Medline Forehead Thermometer, No Touch, 3-3/10"Wx7-1/4"Lx1-2/5"H, WE
SDP
Catalog No. 5000828370
Encompass_Preferred
Forehead thermometer features no touch, infrared technology to quickly measure your patient's temperature while completely eliminating contact. Result indicators help you easily interpret measurement results for accurate readings. Innovative design measures human and surface temperatures in either Celsius or Fahrenheit units. Digital thermometer is approved for both clinical and home settings.
Catalog No. 50-008-28370 Supplier Enterprise Technology Solutions Supplier No. MIIMDSNOTOUCH
Forehead thermometer features no touch, infrared technology to quickly measure your patient's temperature while completely eliminating contact. Result indicators help you easily interpret measurement results for accurate readings. Innovative design measures human and surface temperatures in either Celsius or Fahrenheit units. Digital thermometer is approved for both clinical and home settings.