Learn More
Enterprise Technology Solutions Medline Gauze Bandage Roll, Sterile, 4-1/2"x4 Yards, 100/BX, White
SDP
Catalog No. 5000828437
Encompass_Preferred
Bandage rolls are ideal for bandaging heads, limbs and difficult-to-dress wounds. Six-ply bandages are made of 100 percent cotton gauze. Unique crimped, open-weave design delivers superior absorbency and cushioning. Sterile bandage gently stretches and conforms to body contours. Use as a primary or secondary dressing.
Catalog No. 50-008-28437 Supplier Enterprise Technology Solutions Supplier No. MIIPRM25865
Bandage rolls are ideal for bandaging heads, limbs and difficult-to-dress wounds. Six-ply bandages are made of 100 percent cotton gauze. Unique crimped, open-weave design delivers superior absorbency and cushioning. Sterile bandage gently stretches and conforms to body contours. Use as a primary or secondary dressing.