Learn More
Enterprise Technology Solutions Medline Gauze Sponges, Nonsterile, 4"x4", 8-Ply, 200/BX, White
SDP
Catalog No. 5000828435
Encompass_Preferred
Nonsterile gauze sponges are ideal for wound dressings, wound packing and general wound care. Each latex-free sponge is made of 100 percent woven cotton. Clean, debris-free packing has folded edges to prevent unraveling. 8-ply sponges are also packed in a coated paper envelope to reduce fiber debris. Easy-open envelope is carefully sealed to prevent the intrusion of dust and contaminants.
Catalog No. 50-008-28435 Supplier Enterprise Technology Solutions Supplier No. MIINON25408
Add to Cart
Add to Cart
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Nonsterile gauze sponges are ideal for wound dressings, wound packing and general wound care. Each latex-free sponge is made of 100 percent woven cotton. Clean, debris-free packing has folded edges to prevent unraveling. 8-ply sponges are also packed in a coated paper envelope to reduce fiber debris. Easy-open envelope is carefully sealed to prevent the intrusion of dust and contaminants.