Learn More
Enterprise Technology Solutions Medline Gauze Sponges,Nonwoven,Nonsterile,3"x3",4-Ply,200/BX,White
SDP
Catalog No. 5000827935
Encompass_Preferred
Four-ply, nonsterile gauze sponges are the perfect balance between strength and absorbency compared to traditional cotton gauze. This rayon/polyester-blend gauze produces less lint and is less traumatic to fragile, granulating wound tissue because it's virtually nonadherent. Faster wicking ability promotes a healthier healing environment. Avant Gauze is a standard nonwoven dressing.
Catalog No. 50-008-27935 Supplier Enterprise Technology Solutions Supplier No. MIINON25334
Four-ply, nonsterile gauze sponges are the perfect balance between strength and absorbency compared to traditional cotton gauze. This rayon/polyester-blend gauze produces less lint and is less traumatic to fragile, granulating wound tissue because it's virtually nonadherent. Faster wicking ability promotes a healthier healing environment. Avant Gauze is a standard nonwoven dressing.