Learn More
Enterprise Technology Solutions Medline Gauze Sponges, Sterile, 2"x2",4 Ply, 50/BX, White
SDP
Catalog No. 5000828472
Encompass_Preferred
Four-ply, nonwoven, sterile gauze sponges offer more absorbency and less linting than cotton gauze. Made of a unique rayon/polyester formed fabric, sponges are virtually nonadherent.
Catalog No. 50-008-28472 Supplier Enterprise Technology Solutions Supplier No. MIIPRM21224
Four-ply, nonwoven, sterile gauze sponges offer more absorbency and less linting than cotton gauze. Made of a unique rayon/polyester formed fabric, sponges are virtually nonadherent.