Learn More
Enterprise Technology Solutions Medline Gauze Sponges,Sterile,Cotton Woven, 3"x3", 12 Ply, 80/BX, WE
SDP
Catalog No. 5000828496
Encompass_Preferred
Woven gauze sponges are ideal for wound dressings, wound packing and general wound care. 12-ply, sterile sponges are made of 100 percent cotton. Clean, debris-free packing has folded edges to prevent unraveling. Sponges are also packed in a coated paper envelope to reduce fiber debris. Easy-open envelope is carefully sealed to prevent the intrusion of dust and contaminants.
Catalog No. 50-008-28496 Supplier Enterprise Technology Solutions Supplier No. MIINON21422
Woven gauze sponges are ideal for wound dressings, wound packing and general wound care. 12-ply, sterile sponges are made of 100 percent cotton. Clean, debris-free packing has folded edges to prevent unraveling. Sponges are also packed in a coated paper envelope to reduce fiber debris. Easy-open envelope is carefully sealed to prevent the intrusion of dust and contaminants.