Learn More
Enterprise Technology Solutions Medline Goggles,Lab,Standard,Vent,Elastic Strap,CL
SDP
Catalog No. 5000828453
Encompass_Preferred
Standard Fluid-Protection Lab Goggles feature vents and an elastic strap. They meet ANSI Z87.1 plus standards for impact-resistance to protect your eyes in lab settings. Elastic strap allows a custom fit.
Catalog No. 50-008-28453 Supplier Enterprise Technology Solutions Supplier No. MIINON24776
Standard Fluid-Protection Lab Goggles feature vents and an elastic strap. They meet ANSI Z87.1 plus standards for impact-resistance to protect your eyes in lab settings. Elastic strap allows a custom fit.