Learn More
Enterprise Technology Solutions Medline Nitrile Exam Gloves, Powder-Free, Large, 150/BX, Blue
SDP
Catalog No. 5000828247
Encompass_Preferred
Nitrile exam gloves are the perfect alternative to latex gloves. Full-textured design offers an enhanced grip and is resistant to chemicals. Powder-free, size large gloves are tested for use with chemotherapy drugs.
Catalog No. 50-008-28247 Supplier Enterprise Technology Solutions Supplier No. MIICUR9316
Nitrile exam gloves are the perfect alternative to latex gloves. Full-textured design offers an enhanced grip and is resistant to chemicals. Powder-free, size large gloves are tested for use with chemotherapy drugs.