Learn More
Enterprise Technology Solutions Medline Nitrile Exam Gloves, Powder-Free, Medium, 150/BX, Blue
SDP
Catalog No. 5000828261
Encompass_Preferred
Nitrile exam gloves are the perfect alternative to latex gloves. Full-textured design offers an enhanced grip and is resistant to chemicals. Powder-free, size medium gloves are tested for use with chemotherapy drugs.
Catalog No. 50-008-28261 Supplier Enterprise Technology Solutions Supplier No. MIICUR9315
Nitrile exam gloves are the perfect alternative to latex gloves. Full-textured design offers an enhanced grip and is resistant to chemicals. Powder-free, size medium gloves are tested for use with chemotherapy drugs.