Learn More
Enterprise Technology Solutions Medline Nitrile Exam Gloves, Powder Free, X-Large, 180/BX
SDP
Catalog No. 5000828035
Encompass_Preferred
Aloetouch Ice Exam Gloves offer the comfort of latex in a nitrile glove. These powder-free, latex-free gloves provide great sensitivity as well as dependable protection. The aloe coating helps soothe and protect hardworking hands.
Catalog No. 50-008-28035 Supplier Enterprise Technology Solutions Supplier No. MIIMDS195287
Aloetouch Ice Exam Gloves offer the comfort of latex in a nitrile glove. These powder-free, latex-free gloves provide great sensitivity as well as dependable protection. The aloe coating helps soothe and protect hardworking hands.