Learn More
Enterprise Technology Solutions Medline Nitrile Exam Gloves w/Oatmeal, Large, 250/BX, OFWE
SDP
Supplier:  Enterprise Technology Solutions MIIOAT6803
Encompass_Preferred
Powder-free exam gloves provide extra care for hardworking hands with a new level of skin protection. Colloidal oatmeal helps restore your skin's normal pH and maintain the natural moisture barrier needed to prevent and protect dry skin. Ambidextrous design allows each glove to be used for either hand. Latex-free, nitrile material helps prevent allergic reactions for both home healthcare workers and their clients.
Catalog No. 50-008-28172