Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medline™ Accutouch™ Chemo Nitrile Gloves

Catalog No. 23666025 Shop All Medline Industries Products
Encompass_Preferred
Change view
Click to view available options
Size:
Medium
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
23-666-025 Medium
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-666-025 Supplier Medline™ Supplier No. MDS192085

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Nitrile exam gloves feature tactile sensitivity and high tensile strength for use with chemotherapy drugs.

  • Chemo nitrile exam gloves
  • Powder free
  • Tactile sensitivity and high tensile strength for durable barrier protection
  • Textured for a secure grip
  • Tight cuffs to minimize accidental exposure to blood-borne pathogens and other contaminated fluids
  • Approved for use with chemotherapy drugs, formaldehyde and gluteraldehyde

Specifications

Chemotherapeutic Rated Yes
Applications Healthcare, Laboratory
AQL 0.65
Color Blue
Disposable Single-use
Exterior Finish Fully Textured
Latex Free Yes
Material Nitrile
Powdered Powder Free
Size Medium
Thickness (English) 4.1 mil
Thickness (Metric) 0.10 mm
Length (English) 9.5 in.
Length (Metric) 24.13 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.